Tisk…

3,4-Difluoro-2-methoxybenzyl alcohol 95%

Dodavatel: Apollo Scientific

PC51392-1G PC51392-250MG PC51392-5G
APOSPC51392-1GEA 13860 CZK
APOSPC51392-1G APOSPC51392-250MG APOSPC51392-5G
3,4-Difluoro-2-methoxybenzyl alcohol 95%
3,4-Difluoro-2-methoxybenzyl alcohol
Vzorec: C₈H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28519671
CAS číslo: 1779895-68-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR