Tisk…

4-(Difluoromethoxy)-3-fluorobenzyl bromide ≥95%

Dodavatel: Apollo Scientific

PC51313-250MG PC51313-1G PC51313-5G
APOSPC51313-250MGEA 5920 CZK
APOSPC51313-250MG APOSPC51313-1G APOSPC51313-5G
4-(Difluoromethoxy)-3-fluorobenzyl bromide ≥95%
4-(Difluoromethoxy)-3-fluorobenzyl bromide
Vzorec: C₈H₆BrF₃O
MDL Number: MFCD18394990
CAS číslo: 181806-49-5
UN: 3265

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR