Tisk…

2-Hydroxyethyl trifluoromethyl ether 95%

Dodavatel: Apollo Scientific
Varování

PC5131-1G PC5131-250MG PC5131-500MG
APOSPC5131-1GEA 19440 CZK
APOSPC5131-1G APOSPC5131-250MG APOSPC5131-500MG
2-Hydroxyethyl trifluoromethyl ether 95%
2-Hydroxyethyl trifluoromethyl ether
Vzorec: C₃H₅F₃O₂
Bod varu: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD11858187
CAS číslo: 362631-84-3
UN: 3271
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR