Tisk…

2,4-Difluoro-5-nitrobenzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC51227-1G PC51227-250MG PC51227-5G
APOSPC51227-1GEA 9330 CZK
APOSPC51227-1G APOSPC51227-250MG APOSPC51227-5G
2,4-Difluoro-5-nitrobenzaldehyde 95%
2,4-Difluoro-5-nitrobenzaldehyde
Vzorec: C₇H₃F₂NO₃
MW: 187.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11857779
CAS číslo: 127228-77-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR