Tisk…

Fluoromalonaldehyde, sodium salt

Dodavatel: Apollo Scientific
Varování

Synonyms: Sodium 2-fluoro-3-oxoprop-1-en-1-olate, Fluoropropane-1,3-dial sodium salt

PC51104-1G PC51104-250MG
APOSPC51104-1GEA 9100 CZK
APOSPC51104-1G APOSPC51104-250MG
Fluoromalonaldehyde, sodium salt
Fluoromalonaldehyde, sodium salt
Vzorec: C₃H₂FO₂·Na
Storage Temperature: Ambient
MDL Number: MFCD21362228
CAS číslo: 29548-72-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR