Tisk…

α,α-Difluoro-2-(4-methoxyphenyl)acetic acid

Dodavatel: Apollo Scientific

PC510343-1G PC510343-250MG PC510343-5G
APOSPC510343-1GEA 10650 CZK
APOSPC510343-1G APOSPC510343-250MG APOSPC510343-5G
α,α-Difluoro-2-(4-methoxyphenyl)acetic acid
α,α-Difluoro-2-(4-methoxyphenyl)acetic acid
Vzorec: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11007700
CAS číslo: 1027513-97-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR