Tisk…

Ethyl 2,2-Difluoro-2-(p-tolyl)acetate

Dodavatel: Apollo Scientific

PC510337-1G
APOSPC510337-1GEA 11310 CZK
APOSPC510337-1G
Ethyl 2,2-Difluoro-2-(p-tolyl)acetate
Ethyl 2,2-Difluoro-2-(p-tolyl)acetate
Vzorec: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11007729
CAS číslo: 131323-06-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR