Tisk…

2-(Allyloxy)-1-bromo-4-(trifluoromethoxy)benzene

Dodavatel: Apollo Scientific

PC510151-5G
APOSPC510151-5GEA 31910 CZK
APOSPC510151-5G
2-(Allyloxy)-1-bromo-4-(trifluoromethoxy)benzene
2-(Allyloxy)-1-bromo-4-(trifluoromethoxy)benzene
Vzorec: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12910502
CAS číslo: 1162257-62-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR