Tisk…

2-Ethyl-5-fluorobenzoxazole

Dodavatel: Apollo Scientific

PC510111-5G
APOSPC510111-5GEA 20300 CZK
APOSPC510111-5G
2-Ethyl-5-fluorobenzoxazole
2-Ethyl-5-fluorobenzoxazole
Vzorec: C₉H₈FNO
MW: 165,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19374536
CAS číslo: 1267772-21-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR