Tisk…

(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%

Dodavatel: Apollo Scientific
Nebezpečí

PC5063L-1G PC5063L-250MG
APOSPC5063L-1GEA 5390 CZK
APOSPC5063L-1G APOSPC5063L-250MG
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

For determination of enantiomeric excess of alcohols and amines by NMR.

Vzorec: C₁₀H₈ClF₃O₂
Bod varu: 213…214 °C
Hustota: 1,35 g/cm³ (25 °C)
Bod vzplanutí: 89 °C
Storage Temperature: Freezer
MDL Number: MFCD00044400
CAS číslo: 39637-99-5
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR