Tisk…

4-Ethynyl-3-fluorobenzonitrile 95%

Dodavatel: Apollo Scientific

PC50564-1G PC50564-250MG
APOSPC50564-1GEA 19110 CZK
APOSPC50564-1G APOSPC50564-250MG
4-Ethynyl-3-fluorobenzonitrile 95%
4-Ethynyl-3-fluorobenzonitrile
Vzorec: C₉H₄FN
Storage Temperature: Freezer
CAS číslo: 1581281-30-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR