Tisk…

2-Ethynyl-5-fluorobenzonitrile 95%

Dodavatel: Apollo Scientific

PC50558-1G PC50558-250MG
APOSPC50558-1GEA 17360 CZK
APOSPC50558-1G APOSPC50558-250MG
2-Ethynyl-5-fluorobenzonitrile 95%
2-Ethynyl-5-fluorobenzonitrile
Vzorec: C₉H₄FN
Storage Temperature: Freezer
MDL Number: MFCD18205104
CAS číslo: 1378823-86-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR