Tisk…

2-Bromo-6-iodobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502883-1G PC502883-250MG
APOSPC502883-1GEA 17360 CZK
APOSPC502883-1G APOSPC502883-250MG
2-Bromo-6-iodobenzyl fluoride 95%
2-Bromo-6-iodobenzyl fluoride
Vzorec: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD31634828
CAS číslo: 2244087-75-8
UN: 3261
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR