Tisk…

2,4-Dibromobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502880-1G PC502880-250MG
APOSPC502880-1GEA 17360 CZK
APOSPC502880-1G APOSPC502880-250MG
2,4-Dibromobenzyl fluoride 95%
2,4-Dibromobenzyl fluoride
Vzorec: C₇H₅Br₂F
Storage Temperature: Ambient
MDL Number: MFCD00079786
CAS číslo: 1780254-42-3
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR