Tisk…

2-Amino-4-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Dodavatel: Apollo Scientific

PC502855-1G
APOSPC502855-1GEA 49930 CZK
APOSPC502855-1G
2-Amino-4-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Vzorec: C₁₃H₇F₅INO
Storage Temperature: Ambient
MDL Number: MFCD31630040
CAS číslo: 2244086-23-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR