Tisk…

5-Bromo-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(5-Bromo-2-fluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502777-1G
APOSPC502777-1GEA 33400 CZK
APOSPC502777-1G
5-Bromo-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Vzorec: C₁₃H₅BrF₆O
MW: 371.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31629952
CAS číslo: 2244087-61-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR