Tisk…

1,3,5-Tribromo-2,4,6-trifluorobenzene 97%

Dodavatel: Apollo Scientific

PC502442-1G PC502442-250MG
APOSPC502442-1GEA 2820 CZK
APOSPC502442-1G APOSPC502442-250MG
1,3,5-Tribromo-2,4,6-trifluorobenzene 97%
1,3,5-Tribromo-2,4,6-trifluorobenzene
Vzorec: C₆Br₃F₃
Storage Temperature: Ambient
MDL Number: MFCD00192722
CAS číslo: 2368-49-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR