Tisk…

2,4-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502199-1G PC502199-250MG
APOSPC502199-1GEA 18620 CZK
APOSPC502199-1G APOSPC502199-250MG
2,4-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%
2,4-Difluoro-3-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₄BrF₅O
Storage Temperature: Ambient
MDL Number: MFCD30723444
CAS číslo: 2149591-35-3
UN: 3265
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR