Tisk…

Methyl 3-amino-5-fluoro-4-hydroxybenzoate ≥95%

Dodavatel: Apollo Scientific

PC502069-250MG PC502069-1G PC502069-5G
APOSPC502069-250MGEA 5790 CZK
APOSPC502069-250MG APOSPC502069-1G APOSPC502069-5G
Methyl 3-amino-5-fluoro-4-hydroxybenzoate ≥95%
Methyl 3-amino-5-fluoro-4-hydroxybenzoate
Vzorec: C₈H₈FNO₃
MDL Number: MFCD20725712
CAS číslo: 946121-50-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR