Tisk…

5-Bromo-2,4-difluorotoluene

Dodavatel: Apollo Scientific

PC501997-1G PC501997-25G PC501997-5G
APOSPC501997-1GEA 622 CZK
APOSPC501997-1G APOSPC501997-25G APOSPC501997-5G
5-Bromo-2,4-difluorotoluene
5-Bromo-2,4-difluorotoluene
Vzorec: C₇H₅BrF₂
Storage Temperature: Ambient
MDL Number: MFCD15144620
CAS číslo: 159277-47-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR