Tisk…

Methyl-5-bromo-2,3-difluorobenzoate

Dodavatel: Apollo Scientific
Varování

PC501961-1G PC501961-250MG PC501961-5G
APOSPC501961-1GEA 2980 CZK
APOSPC501961-1G APOSPC501961-250MG APOSPC501961-5G
Methyl-5-bromo-2,3-difluorobenzoate
Methyl-5-bromo-2,3-difluorobenzoate
Vzorec: C₈H₅BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12026006
CAS číslo: 1150163-69-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR