Tisk…

Methyl 4-fluoro-3-(hydroxymethyl)benzoate 95%

Dodavatel: Apollo Scientific
Varování

PC501655-100MG PC501655-1G PC501655-250MG
APOSPC501655-100MGEA 9030 CZK
APOSPC501655-100MG APOSPC501655-1G APOSPC501655-250MG
Methyl 4-fluoro-3-(hydroxymethyl)benzoate 95%
Methyl 4-fluoro-3-(hydroxymethyl)benzoate
Vzorec: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD15527250
CAS číslo: 816449-69-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR