Tisk…

3-Fluoro-2-nitrobenzotrifluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC501476-1G PC501476-5G PC501476-250MG
APOSPC501476-1GEA 3110 CZK
APOSPC501476-1G APOSPC501476-5G APOSPC501476-250MG
3-Fluoro-2-nitrobenzotrifluoride 95%
3-Fluoro-2-nitrobenzotrifluoride
Vzorec: C₇H₃F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD12032180
CAS číslo: 1214335-98-4
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR