Tisk…

2-Bromo-4-chloro-6-fluorobenzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

PC50135-1G PC50135-250MG
APOSPC50135-1GEA 18620 CZK
APOSPC50135-1G APOSPC50135-250MG
2-Bromo-4-chloro-6-fluorobenzyl alcohol 95%
2-Bromo-4-chloro-6-fluorobenzyl alcohol
Vzorec: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD24848700
CAS číslo: 1449008-26-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR