Tisk…

4-Bromo-2-fluoro-3-methylaniline 95%

Dodavatel: Apollo Scientific
Varování

PC501223-1G PC501223-250MG PC501223-5G
APOSPC501223-1GEA 3020 CZK
APOSPC501223-1G APOSPC501223-250MG APOSPC501223-5G
4-Bromo-2-fluoro-3-methylaniline 95%
4-Bromo-2-fluoro-3-methylaniline
Vzorec: C₇H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD26687842
CAS číslo: 1540204-53-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR