Tisk…

3-Fluoro-4-(trifluoromethoxy)phenacyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC501202-1G PC501202-250MG PC501202-5G
APOSPC501202-1GEA 17990 CZK
APOSPC501202-1G APOSPC501202-250MG APOSPC501202-5G
3-Fluoro-4-(trifluoromethoxy)phenacyl bromide 95%
3-Fluoro-4-(trifluoromethoxy)phenacyl bromide
Vzorec: C₉H₅BrF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD28348503
CAS číslo: 1448246-57-8
UN: 3261
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR