Tisk…

Lithium tetrafluoroborate, anhydrous 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC5010-100G PC5010-25G
APOSPC5010-100GEA 2660 CZK
APOSPC5010-100G APOSPC5010-25G
Lithium tetrafluoroborate, anhydrous 98%
Lithium tetrafluoroborate
Vzorec: BF₄Li
Bod tání: 309…311 °C
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS číslo: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR