Tisk…

2-(2,2,2-Trifluoroethoxy)benzoic acid

Dodavatel: Apollo Scientific

PC500832-1G PC500832-5G
APOSPC500832-1GEA 8140 CZK
APOSPC500832-1G APOSPC500832-5G
2-(2,2,2-Trifluoroethoxy)benzoic acid
2-(2,2,2-Trifluoroethoxy)benzoic acid
Vzorec: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD08556302
CAS číslo: 35480-46-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR