Tisk…

Potassium methyltrifluoroborate 97%

Dodavatel: Apollo Scientific
Varování

PC50060-10G PC50060-5G
APOSPC50060-10GEA 2940 CZK
APOSPC50060-10G APOSPC50060-5G
Potassium methyltrifluoroborate 97%
Potassium methyltrifluoroborate
Vzorec: CH₃BF₃K
Bod tání: 168…183 °C
Storage Temperature: Ambient
MDL Number: MFCD08276757
CAS číslo: 13862-28-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR