Tisk…

5-Bromo-2-fluorophenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC500502-1G PC500502-250MG PC500502-5G
APOSPC500502-1GEA 11580 CZK
APOSPC500502-1G APOSPC500502-250MG APOSPC500502-5G
5-Bromo-2-fluorophenacyl bromide
5-Bromo-2-fluorophenacyl bromide
Vzorec: C₈H₅Br₂FO
Storage Temperature: Refrigerator
MDL Number: MFCD23711739
CAS číslo: 1427413-65-7
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR