Tisk…

3-Fluoro-4-nitrobenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC500352-1G PC500352-25G PC500352-5G
APOSPC500352-1GEA 1160 CZK
APOSPC500352-1G APOSPC500352-25G APOSPC500352-5G
3-Fluoro-4-nitrobenzenesulphonyl chloride
3-Fluoro-4-nitrobenzenesulphonyl chloride
Vzorec: C₆H₃ClFNO₄S
Storage Temperature: Ambient
MDL Number: MFCD03412240
CAS číslo: 86156-93-6
UN: 3261
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR