Tisk…

2-Amino-4-bromo-3-fluorobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC500189-5G PC500189-1G PC500189-250MG
APOSPC500189-5GEA 11910 CZK
APOSPC500189-5G APOSPC500189-1G APOSPC500189-250MG
2-Amino-4-bromo-3-fluorobenzoic acid 95%
2-Amino-4-bromo-3-fluorobenzoic acid
Vzorec: C₇H₅BrFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD27664889
CAS číslo: 1416013-62-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR