Tisk…

2-Fluoro-4-methoxybenzotrifluoride

Dodavatel: Apollo Scientific

PC499008-1G PC499008-5G
APOSPC499008-1GEA 6250 CZK
APOSPC499008-1G APOSPC499008-5G
2-Fluoro-4-methoxybenzotrifluoride
2-Fluoro-4-methoxybenzotrifluoride
Vzorec: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185641
CAS číslo: 1214344-33-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR