Tisk…

2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2-(Difluoromethoxy)-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC49715-1G PC49715-250MG PC49715-5G
APOSPC49715-1GEA 27970 CZK
APOSPC49715-1G APOSPC49715-250MG APOSPC49715-5G
2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester 95%
2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester
Vzorec: C₁₃H₁₆BF₃O₃
MW: 288.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30486443
CAS číslo: 1417301-27-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR