Tisk…

4-Fluoro-3-iodoaniline

Dodavatel: Apollo Scientific
Varování

PC49605-1G PC49605-250MG PC49605-5G
APOSPC49605-1GEA 3480 CZK
APOSPC49605-1G APOSPC49605-250MG APOSPC49605-5G
4-Fluoro-3-iodoaniline
4-Fluoro-3-iodoaniline
Vzorec: C₆H₅FIN
Storage Temperature: Ambient
MDL Number: MFCD18396914
CAS číslo: 647025-62-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR