Tisk…

4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine oxalate

Dodavatel: Apollo Scientific
Varování

PC49515-1G PC49515-250MG
APOSPC49515-1GEA 42650 CZK
APOSPC49515-1G APOSPC49515-250MG
4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine oxalate
4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine oxalate
Vzorec: C₉H₁₄F₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD18909670
CAS číslo: 1298094-30-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR