Tisk…

4-(Trifluoroacetyl)piperidine, N-BOC protected 95%

Dodavatel: Apollo Scientific

PC49510-1G PC49510-250MG
APOSPC49510-1GEA 7670 CZK
APOSPC49510-1G APOSPC49510-250MG
4-(Trifluoroacetyl)piperidine, N-BOC protected 95%
4-(Trifluoroacetyl)piperidine, N-BOC protected
Vzorec: C₁₂H₁₈F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD11501650
CAS číslo: 1093759-80-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR