Tisk…

2-Fluoro-4-iodonicotinic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-4-iodo-nicotinic acid

PC49358-1G
APOSPC49358-1GEA 622 CZK
APOSPC49358-1G
2-Fluoro-4-iodonicotinic acid
2-Fluoro-4-iodonicotinic acid
Vzorec: C₆H₃FINO₂
MW: 267.0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095310
CAS číslo: 884494-51-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR