Tisk…

3-(Difluoromethyl)benzonitrile

Dodavatel: Apollo Scientific
Varování

PC49196-1G PC49196-5G PC49196-25G
APOSPC49196-1GEA 744 CZK
APOSPC49196-1G APOSPC49196-5G APOSPC49196-25G
3-(Difluoromethyl)benzonitrile
3-(Difluoromethyl)benzonitrile
Vzorec: C₈H₅F₂N
MW: 153.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19053256
CAS číslo: 55805-13-5
UN: 3276
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR