Tisk…

4-Methoxy-2-(trifluoromethoxy)benzaldehyde

Dodavatel: Apollo Scientific
Varování

PC49158-10G PC49158-5G
APOSPC49158-10GEA 8700 CZK
APOSPC49158-10G APOSPC49158-5G
4-Methoxy-2-(trifluoromethoxy)benzaldehyde
4-Methoxy-2-(trifluoromethoxy)benzaldehyde
Vzorec: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD06660317
CAS číslo: 886503-52-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR