Tisk…

2,4-Difluoro-3-methylbenzaldehyde 95%

Dodavatel: Apollo Scientific

PC48740-1G PC48740-5G
APOSPC48740-1GEA 2530 CZK
APOSPC48740-1G APOSPC48740-5G
2,4-Difluoro-3-methylbenzaldehyde 95%
2,4-Difluoro-3-methylbenzaldehyde
Vzorec: C₈H₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD07777152
CAS číslo: 847502-88-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR