Tisk…

2-(Difluoromethoxy)benzeneboronic acid, pinacol ester

Dodavatel: Apollo Scientific
Varování

PC48609-1G PC48609-250MG PC48609-5G
APOSPC48609-1GEA 2610 CZK
APOSPC48609-1G APOSPC48609-250MG APOSPC48609-5G
2-(Difluoromethoxy)benzeneboronic acid, pinacol ester
2-(Difluoromethoxy)benzeneboronic acid, pinacol ester
Vzorec: C₁₃H₁₇BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD16996403
CAS číslo: 960067-33-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR