Tisk…

[4-(Difluoromethyl)phenyl]methanol

Dodavatel: Apollo Scientific
Varování

PC48346-250MG
APOSPC48346-250MGEA 5260 CZK
APOSPC48346-250MG
[4-(Difluoromethyl)phenyl]methanol
[4-(Difluoromethyl)phenyl]methanol
Vzorec: C₈H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD20488855
CAS číslo: 444915-77-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR