Tisk…

(4-Chlorooctafluorobutyl)trifluorooxirane

Dodavatel: Apollo Scientific
Varování

PC450565-1G PC450565-25G PC450565-5G
APOSPC450565-1GEA 1160 CZK
APOSPC450565-1G APOSPC450565-25G APOSPC450565-5G
(4-Chlorooctafluorobutyl)trifluorooxirane
(4-Chlorooctafluorobutyl)trifluorooxirane
Vzorec: C₆ClF₁₁O
MW: 332.499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619622
CAS číslo: 50838-69-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR