Tisk…

Bis[3-(trifluoromethyl)phenyl] sulphate

Dodavatel: Apollo Scientific

PC450078-1G PC450078-250MG
APOSPC450078-1GEA 8100 CZK
APOSPC450078-1G APOSPC450078-250MG
Bis[3-(trifluoromethyl)phenyl] sulphate
Bis[3-(trifluoromethyl)phenyl] sulphate
Vzorec: C₁₄H₈F₆O₄S
Storage Temperature: Ambient
MDL Number: MFCD24842696
CAS číslo: 1417220-66-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR