Tisk…

Allyl 2,2-difluoroethyl ether

Dodavatel: Apollo Scientific

PC450065-1G
APOSPC450065-1GEA 8100 CZK
APOSPC450065-1G
Allyl 2,2-difluoroethyl ether
Allyl 2,2-difluoroethyl ether
Vzorec: C₅H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD19442068
CAS číslo: 111512-41-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR