Tisk…

(E)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile

Dodavatel: Apollo Scientific

PC450005-100MG
APOSPC450005-100MGEA 8100 CZK
APOSPC450005-100MG
(E)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile
(E)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile
Vzorec: C₁₀H₈FN
Storage Temperature: Ambient
MDL Number: MFCD22124012
CAS číslo: 1365836-27-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR