Tisk…

2-[4-(Chloromethyl)phenyl]-5-fluoropyrimidine, tech

Dodavatel: Apollo Scientific

PC448205-250MG
APOSPC448205-250MGEA 21390 CZK
APOSPC448205-250MG
2-[4-(Chloromethyl)phenyl]-5-fluoropyrimidine, tech
2-[4-(Chloromethyl)phenyl]-5-fluoropyrimidine
Vzorec: C₁₁H₈ClFN₂
Storage Temperature: Ambient
MDL Number: MFCD23160098
CAS číslo: 1139432-30-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR