Tisk…

2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene 98%

Dodavatel: Apollo Scientific

PC448035-5G
APOSPC448035-5GEA 2320 CZK
APOSPC448035-5G
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene 98%
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene
Vzorec: C₁₈H₁₄BrFS
MW: 361.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21496340
CAS číslo: 1030825-20-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR