Tisk…

3-Fluoro-5-(trifluoromethyl)phenol 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC4376P-10G PC4376P-50G PC4376P-1G PC4376P-5G PC4376P-25G
APOSPC4376P-10GEA 0 CZK
APOSPC4376P-10G APOSPC4376P-50G APOSPC4376P-1G APOSPC4376P-5G APOSPC4376P-25G
3-Fluoro-5-(trifluoromethyl)phenol 98%
3-Fluoro-5-(trifluoromethyl)phenol
Vzorec: C₇H₄F₄O
Storage Temperature: Ambient
MDL Number: MFCD00061288
CAS číslo: 172333-87-8
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR